2-Bromo-5-hydroxybenzaldehyde ≥97%

Supplier: Matrix Scientific

007674-5G 007674-25G
101793-212EA 36.92 USD
101793-212 101793-214
2-Bromo-5-hydroxybenzaldehyde ≥97%
2-Bromo-5-hydroxybenzaldehyde

Hazard code(s): Irritant, Store in Freezer

Formula: C₇H₅BrO₂
MW: 201.02 g/mol
MDL Number: MFCD03033065
CAS Number: 2973-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR