5-Bromopyrimidine ≥98%

Supplier: Matrix Scientific
Warning

007659-25G 007659-100G
101793-194EA 107.69 USD
101793-194 101793-196
5-Bromopyrimidine ≥98%
5-Bromopyrimidine

Hazard code(s): Irritant, Refrigerate

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Boiling Pt: 168 °C (727 -rr)
Melting Pt: 75 - 76 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1

Order Now

Specification Test Results

Melting point 69-72°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR