8-Quinolinecarboxylic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Quinoline-8-carboxylic acid
007648-1G
007648-5G
101793-178EA
43.08
USD
101793-178
101793-180
8-Quinolinecarboxylic acid ≥98%
8-Quinolinecarboxylic acid
Hazard code(s): Irritant
Formula:
C₁₀H₇NO₂ MW: 173.17 g/mol Melting Pt: 187 °C |
MDL Number:
MFCD00047619 CAS Number: 86-59-9 |
Specification Test Results
Melting point | 187°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...