1,4-Phenylenediacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2'-(1,4-Phenylene)diacetic acid

007638-1G 007638-5G 007638-25G
101793-158EA 29.23 USD
101793-158 101793-160 101793-162
1,4-Phenylenediacetic acid ≥95%
1,4-Phenylenediacetic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 249…251 °C
MDL Number: MFCD00004354
CAS Number: 7325-46-4
EINECS: 230-801-2

Order Now

Specification Test Results

Melting point 249-251°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR