4-Chlorobutyl phenyl ether ≥98%

Supplier: Matrix Scientific

007636-5G 007636-25G
101793-150EA 38.46 USD
101793-150 101793-152
4-Chlorobutyl phenyl ether ≥98%
4-Chlorobutyl phenyl ether

Hazard code(s): Irritant

Formula: (C₆H₅)O(CH₂)₄Cl
MW: 184.67 g/mol
Boiling Pt: 147 °C ( 12mm)
Melting Pt: 20 °C
Density: 1.09 g/cm³
MDL Number: MFCD00001012
CAS Number: 2651-46-9
EINECS: 220-174-3

Order Now

Specification Test Results

Boiling point 147°C/ 12mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR