3-Nitro-p-anisic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methoxy-3-nitrobenzoic acid

007615-25G 007615-100G
101793-120EA 47.69 USD
101793-120 101793-122
3-Nitro-p-anisic acid ≥97%
3-Nitro-p-anisic acid

Hazard code(s): Irritant

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Boiling Pt: 372 °C (1013 hPa)
Melting Pt: 185 °C
MDL Number: MFCD00007256
CAS Number: 89-41-8
EINECS: 201-906-0

Order Now

Specification Test Results

Melting point 185-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR