2-Methoxy-p-toluic acid ≥98%

Supplier: Matrix Scientific

007613-1G 007613-5G
101793-112EA 21.54 USD
101793-112 101793-114
2-Methoxy-p-toluic acid ≥98%
2-Methoxy-p-toluic acid

Hazard code(s): Irritant

Formula: C₉H₁₀O₃
Melting Pt: 76 °C
MDL Number: MFCD00020380
CAS Number: 704-45-0

Order Now

Specification Test Results

Melting point 76°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR