(2-Iodophenyl)acetic acid ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 2-(2-Iodophenyl)acetic acid

007606-5G 007606-25G
101793-104EA 38.46 USD
101793-104 101793-106
(2-Iodophenyl)acetic acid ≥98% for synthesis
(2-Iodophenyl)acetic acid

Hazard code(s): Irritant, Light Sensitive

Formula: (C₆H₄I)CH₂COOH
Melting Pt: 115…117 °C
MDL Number: MFCD00046546
CAS Number: 18698-96-9

Order Now

Specification Test Results

Melting point 115-117°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR