1-Iodo-4-nitrobenzene ≥98%

Supplier: Matrix Scientific

007605-25G 007605-100G
101793-100EA 47.69 USD
101793-100 101793-102
1-Iodo-4-nitrobenzene ≥98%
1-Iodo-4-nitrobenzene

Hazard code(s): Irritant, Store in Freezer, Light Sensitive

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 289 °C (772
Melting Pt: 171…173 °C
Density: 2.273 g/cm³ (20 °C)
MDL Number: MFCD00007299
CAS Number: 636-98-6
EINECS: 211-272-7

Order Now

Specification Test Results

Boiling point 289°C/772
Melting point 171-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR