7-Quinolinol ≥97%

Supplier: Matrix Scientific

Synonyms: Quinolin-7-ol, 7-Hydroxyquinoline

007603-1G 007603-5G
101793-092EA 24.62 USD
101793-092 101793-094
7-Quinolinol ≥97%
7-Quinolinol

Hazard code(s): Irritant

Formula: C₉H₇NO
MW: 145.16 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 239 °C
MDL Number: MFCD00016730
CAS Number: 580-20-1
EINECS: 209-457-2

Order Now

Specification Test Results

Melting point 239°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR