4-Hydroxy-3-nitrobenzoic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
007601-25G
007601-100G
101793-086EA
60
USD
101793-086
101793-088
4-Hydroxy-3-nitrobenzoic acid ≥98%
4-Hydroxy-3-nitrobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₅NO₅ MW: 183.12 g/mol Melting Pt: 182 - 185 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007119 CAS Number: 616-82-0 EINECS: 210-494-1 |
Specification Test Results
Melting point | 182-184°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...