1,4-Dibenzoylbutane ≥97%

Supplier: Matrix Scientific

007546-5G 007546-25G
101793-014EA 76.92 USD
101793-014 101793-016
1,4-Dibenzoylbutane ≥97%
1,4-Dibenzoylbutane

Hazard code(s): Irritant

Formula: C₁₈H₁₈O₂
Melting Pt: 107 °C
MDL Number: MFCD00004761
CAS Number: 3375-38-0
EINECS: 222-166-5

Order Now

Specification Test Results

Melting point 107°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR