3-Phenylbenzyl bromide ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Bromomethyl)biphenyl, m-(Bromomethyl)biphenyl

007522-1G 007522-5G
101792-978EA 53.85 USD
101792-978 101792-980
3-Phenylbenzyl bromide ≥97%
3-Phenylbenzyl bromide

Hazard code(s): Irritant, Lachrymator

Formula: C₁₃H₁₁Br
MW: 247.13 g/mol
Boiling Pt: 185 °C (12mm)
Melting Pt: 43 °C
MDL Number: MFCD03452790
CAS Number: 14704-31-5

Order Now

Specification Test Results

Boiling point 185°C/12mm
Melting point 43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR