4-Nitroanthranilic acid ≥95%
Supplier: Matrix Scientific
|
Warning
|
007500-25G
007500-100G
101792-936EA
43.08
USD
101792-936
101792-938
4-Nitroanthranilic acid ≥95%
4-Nitroanthranilic acid
Hazard code(s): Irritant
Formula:
C₇H₆N₂O₄ Melting Pt: 268 °C (dec.) |
MDL Number:
MFCD00007262 CAS Number: 619-17-0 EINECS: 210-583-5 |
Specification Test Results
Melting point | 268°C(dec) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...