6-Aminohexan-1-ol ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 6-Amino-1-hexanol

007496-5G 007496-25G
101792-930EA 29.23 USD
101792-930 101792-932
6-Aminohexan-1-ol ≥97%
6-Aminohexan-1-ol

Hazard code(s): Irritant, Hygroscopic

Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 136…139 °C (30mm)
Melting Pt: 57…59 °C
MDL Number: MFCD00008241
CAS Number: 4048-33-3
EINECS: 223-748-1

Order Now

Specification Test Results

Boiling point 136-139°C/3
Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR