3-Iodobenzaldehyde ≥97%

Supplier: Matrix Scientific

007491-1G 007491-5G
101792-918EA 29.23 USD
101792-918 101792-920
3-Iodobenzaldehyde ≥97%
3-Iodobenzaldehyde

Hazard code(s): Irritant, Store in Freezer, Air Sensitive

Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 124…125 °C (13mm)
Melting Pt: 55…57 °C
MDL Number: MFCD00039573
CAS Number: 696-41-3

Order Now

Specification Test Results

Boiling point 124-125°C/1
Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR