2-Iodobenzaldehyde ≥98%

Supplier: Matrix Scientific
Warning

007490-1G 007490-5G 007490-25G
101792-912EA 43.08 USD
101792-912 101792-914 101792-916
2-Iodobenzaldehyde ≥98%
2-Iodobenzaldehyde

Hazard code(s): Irritant, Store in Freezer, Air Sensitive

Formula: C₇H₅IO
MW: 232.02 g/mol
Boiling Pt: 102 °C (3mm)
Melting Pt: 36…37 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039570
CAS Number: 26260-02-6

Order Now

Specification Test Results

Boiling point 102°C/3mm
Melting point 36-37°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR