(3-Iodophenyl)acetic acid ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 2-(3-Iodophenyl)acetic acid

007466-1G 007466-5G
101792-846EA 69.23 USD
101792-846 101792-848
(3-Iodophenyl)acetic acid ≥97% for synthesis
(3-Iodophenyl)acetic acid

Hazard code(s): Irritant, Light Sensitive

Formula: (C₆H₄I)CH₂COOH
Melting Pt: 129…131 °C
MDL Number: MFCD00046548
CAS Number: 1878-69-9

Order Now

Specification Test Results

Melting point 129-131°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR