3-(4-Ethoxyphenyl)propionic acid ≥98%

Supplier: Matrix Scientific

007463-5G 007463-25G
101792-838EA 81.54 USD
101792-838 101792-840
3-(4-Ethoxyphenyl)propionic acid ≥98%
3-(4-Ethoxyphenyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₁H₁₄O₃
Melting Pt: 102…104 °C
MDL Number: MFCD00016557
CAS Number: 4919-34-0

Order Now

Specification Test Results

Melting point 102-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR