4-Amino-m-toluic acid ≥97%

Supplier: Matrix Scientific
Warning

007452-5G 007452-25G 007452-100G
101792-802EA 18.46 USD
101792-802 101792-804 102663-568
4-Amino-m-toluic acid ≥97%
4-Amino-m-toluic acid

Hazard code(s): Irritant

Formula: C₈H₉NO₂
Boiling Pt: 340 °C (1013 hPa)
Melting Pt: 169…171 °C
MDL Number: MFCD00007736
CAS Number: 2486-70-6
EINECS: 219-629-9

Order Now

Specification Test Results

Melting point 169-171°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR