2-(Trifluoromethyl)benzimidazole ≥98%
Supplier: Matrix Scientific
Synonyms:
2-(Trifluoromethyl)-1H-benzimidazole, 2-(Trifluoromethyl)-1H-benzo[d]imidazole, TFB-2
007447-5G
007447-25G
101792-786EA
44.62
USD
101792-786
101792-788
2-(Trifluoromethyl)benzimidazole ≥98%
2-(Trifluoromethyl)benzimidazole
Hazard code(s): Irritant
Formula:
C₈H₅F₃N₂ MW: 186.14 g/mol Boiling Pt: 341 °C (1013 hPa) Melting Pt: 210 °C Density: 1.487 g/cm³ (20 °C) |
MDL Number:
MFCD00022675 CAS Number: 312-73-2 UN: 2811 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...