1,2-Dibromotetrafluorobenzene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 1,2-Dibromo-3,4,5,6-tetrafluorobenzene

007422-5G 007422-25G
101792-716EA 107.69 USD
101792-716 101792-718
1,2-Dibromotetrafluorobenzene ≥99%
1,2-Dibromotetrafluorobenzene

Hazard code(s): Irritant

Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 198 °C
Melting Pt: 12 °C
Density: 2.23 g/cm³ (20 °C)
MDL Number: MFCD00039209
CAS Number: 827-08-7
EINECS: 212-564-7

Order Now

Specification Test Results

Boiling point 198°C
Density 2.235
ND20 1.5159

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR