2,3-Difluorophenylboronic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Dihydroxy(2,3-difluorophenyl)borane, 2,3-Difluorobenzeneboronic acid, (2,3-Difluorophenyl)metaboric acid

007406-5G 007406-25G
101792-678EA 23.08 USD
101792-678 101792-680
2,3-Difluorophenylboronic acid ≥98%
2,3-Difluorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Melting Pt: ∼225 °C (dec.)
MDL Number: MFCD01863170
CAS Number: 121219-16-7

Order Now

Specification Test Results

Melting point ca 225°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR