3,5-Dichlorophenylboronic acid ≥98%

Supplier: Matrix Scientific
Warning

007405-5G 007405-25G 007405-100G
101792-672EA 21.54 USD
101792-672 101792-674 101792-676
3,5-Dichlorophenylboronic acid ≥98%
3,5-Dichlorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: ∼315 °C (dec.)
MDL Number: MFCD00051935
CAS Number: 67492-50-6

Order Now

Specification Test Results

Melting point ca 315°C(de

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR