3,4-Dichlorophenylboronic acid ≥97%

Supplier: Matrix Scientific
Warning

007404-5G 007404-25G
101792-668EA 38.46 USD
101792-668 101792-670
3,4-Dichlorophenylboronic acid ≥97%
3,4-Dichlorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 270…274 °C
MDL Number: MFCD01074646
CAS Number: 151169-75-4

Order Now

Specification Test Results

Melting point 270-274°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR