5-Bromo-2-thiopheneboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-thienylboronic acid, 5-Bromothiophene-2-boronic acid, (5-Bromothiophen-2-yl)boronic acid

007402-1G 007402-5G
101792-662EA 35.38 USD
101792-662 101792-664
5-Bromo-2-thiopheneboronic acid ≥97%
5-Bromo-2-thiopheneboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₄H₄BBrO₂S
MW: 206.85 g/mol
MDL Number: MFCD02094028
CAS Number: 162607-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR