3-Bromophenylboronic acid ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: Dihydroxy(3-bromophenyl)borane, (3-Bromophenyl)metaboric acid, (3-Bromophenyl)boronic acid, 3-Bromobenzeneboronic acid

007401-5G 007401-25G 007401-100G
101792-656EA 36.92 USD
101792-656 101792-658 101792-660
3-Bromophenylboronic acid ≥98% for synthesis
3-Bromophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₆BBrO₂
Melting Pt: 164…168 °C
MDL Number: MFCD00239386
CAS Number: 89598-96-9

Order Now

Specification Test Results

Melting point 164-168°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR