2,4,6-Trimethylphenylboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane

007398-5G 007398-25G
101792-644EA 38.46 USD
101792-644 102832-788
2,4,6-Trimethylphenylboronic acid ≥95%
2,4,6-Trimethylphenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD00236060
CAS Number: 5980-97-2

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR