3-(Methylthio)phenylboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Thioanisoleboronic acid, 3-(Methylthio)benzeneboronic acid
007392-5G
007392-25G
101792-638EA
29.23
USD
101792-638
101792-640
3-(Methylthio)phenylboronic acid ≥97%
3-(Methylthio)phenylboronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₇H₉BO₂S MW: 168.02 g/mol Melting Pt: 155…157 °C |
MDL Number:
MFCD01319105 CAS Number: 128312-11-8 |
Specification Test Results
Melting point | 155-157°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...