1,4-Phenyldiboronic acid ≥99%

Supplier: Matrix Scientific

Synonyms: p-Phenylenebisboronic acid, 1,4-Phenylenediboronic acid, 1,4-Phenyldiboric acid, 1,4-Benzenediboronic acid

007386-1G 007386-5G 007386-25G
101792-620EA 29.23 USD
101792-620 101792-622 101792-624
1,4-Phenyldiboronic acid ≥99%
1,4-Phenyldiboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₈B₂O₄
MW: 165.75 g/mol
MDL Number: MFCD00236018
CAS Number: 4612-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR