2-Fluoro-3-methoxyphenylboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid
007381-5G
007381-25G
101792-604EA
32.31
USD
101792-604
101792-606
2-Fluoro-3-methoxyphenylboronic acid ≥97%
2-Fluoro-3-methoxyphenylboronic acid
Hazard code(s): Irritant, Refrigerate
Formula:
C₇H₈BFO₃ MW: 169.95 g/mol |
MDL Number:
MFCD02094728 CAS Number: 352303-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...