3,5-Difluorophenylboronic acid ≥98%

Supplier: Matrix Scientific
Warning

007378-25G 007378-100G
101792-598EA 73.85 USD
101792-598 101792-600
3,5-Difluorophenylboronic acid ≥98%
3,5-Difluorophenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅BF₂O₂
Melting Pt: >300 °C
MDL Number: MFCD01318138
CAS Number: 156545-07-2

Order Now

Specification Test Results

Melting point >300°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR