4-Hydroxybenzamidine hydrochloride ≥95%

Supplier: Matrix Scientific

007340-1G 007340-5G
101792-492EA 43.08 USD
101792-492 101792-494
4-Hydroxybenzamidine hydrochloride ≥95%
4-Hydroxybenzamidine hydrochloride

Hazard code(s): Irritant

Formula: C₇H₉ClN₂O
Melting Pt: 223…226 °C
MDL Number: MFCD00136405
CAS Number: 38148-63-9

Order Now

Specification Test Results

Melting point 223-226°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR