4-Chloro-2-fluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

007252-25G 007252-100G
101792-254EA 29.23 USD
101792-254 101792-256
4-Chloro-2-fluorobenzoic acid ≥98%
4-Chloro-2-fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄ClFO₂
Melting Pt: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00042468
CAS Number: 446-30-0
EINECS: 207-162-3

Order Now

Specification Test Results

Melting point 206-208°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR