3,5-Dichlorophenylhydrazine ≥98%

Supplier: Matrix Scientific

007245-5G 007245-25G
101792-240EA 66.15 USD
101792-240 101792-242
3,5-Dichlorophenylhydrazine ≥98%
3,5-Dichlorophenylhydrazine

Hazard code(s): Irritant-Harmful, Refrigerate

Formula: C₆H₆Cl₂N₂
MW: 177.03
Melting Pt: 111…114 °C
MDL Number: MFCD00051527
CAS Number: 39943-56-1
UN: 2811
ADR: 6.1 I

Order Now

Specification Test Results

Melting point 111-114°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR