4-Chloronicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-pyridinecarboxylic acid, 4-Chloropyridine-3-carboxylic acid

007241-5G 007241-25G
101792-236EA 41.54 USD
101792-236 101792-238
4-Chloronicotinic acid ≥95%
4-Chloronicotinic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 301 °C
Melting Pt: 139…143 °C
Density: 1.47 g/cm³
MDL Number: MFCD00128860
CAS Number: 10177-29-4

Order Now

Specification Test Results

Melting point 200-215°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR