2-Chloro-6-methylpyridine ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 6-Chloro-2-picoline

007240-5G 007240-25G
101792-232EA 23.08 USD
101792-232 101792-234
2-Chloro-6-methylpyridine ≥97% for synthesis
2-Chloro-6-methylpyridine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 64
Density: 1.167 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006245
CAS Number: 18368-63-3
EINECS: 242-241-6
UN: 2810
ADR: 6.1 III

Order Now

Specification Test Results

Boiling point 64-68°C/10m
Density 1.167
ND20 15,265


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR