6-Fluoro-1-indanone ≥98%
Supplier: Matrix Scientific
|
Warning
|
007215-5G
007215-25G
101792-208EA
29.23
USD
101792-208
101792-210
6-Fluoro-1-indanone ≥98%
6-Fluoro-1-indanone
Hazard code(s): Irritant
Formula:
C₉H₇FO MW: 150.15 g/mol Boiling Pt: 60 °C (0.5mm) Melting Pt: 53…55 °C |
MDL Number:
MFCD01318147 CAS Number: 1481-32-9 |
Specification Test Results
Boiling point | 60°C/0.5mm |
Melting point | 53-55°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...