2-Fluorothioanisole ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 1-Fluoro-2-(methylthio)benzene, 2-Fluorophenyl methyl sulphide

007210-25G 007210-100G
101792-196EA 113.85 USD
101792-196 101792-198
2-Fluorothioanisole ≥98%
2-Fluorothioanisole

Hazard code(s): Irritant, Stench

Formula: C₇H₇FS
MW: 142.2 g/mol
Boiling Pt: 197…199 °C
MDL Number: MFCD03093762
CAS Number: 655-20-9

Order Now

Specification Test Results

Boiling point 197-199°C
ND20 1.559

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR