6-Chloropicolinic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Chloropyridine-2-carboxylic acid, 2-Chloropyridine-6-carboxylic acid
007202-5G
007202-25G
007202-100G
101792-182EA
15.38
USD
101792-182
101792-184
102832-786
6-Chloropicolinic acid ≥97%
6-Chloropicolinic acid
Hazard code(s): Irritant
Formula:
C₆H₄ClNO₂ MW: 157.56 g/mol Melting Pt: 190…191 °C |
MDL Number:
MFCD00155390 CAS Number: 4684-94-0 |
Specification Test Results
Melting point | 190-191°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...