1,1,2-Trichloro-2,2-difluoroethane ≥99% for synthesis

Supplier: Matrix Scientific

Synonyms: 1,2,2-Trichloro-1,1-difluoroethane, HCFC-122, 1,1-Difluoro-1,2,2-trichloroethane

007198-5G 007198-25G
101792-174EA 63.08 USD
101792-174 101792-176
1,1,2-Trichloro-2,2-difluoroethane ≥99% for synthesis
1,1,2-Trichloro-2,2-difluoroethane

Hazard code(s): Ozone Depleter, Irritant

Formula: Cl₂HCCClF₂
MW: 169.38
Boiling Pt: 72 °C
Melting Pt: –141 °C
Density: 1.559 g/cm³
MDL Number: MFCD00039339
CAS Number: 354-21-2
EINECS: 206-548-9

Order Now

Specification Test Results

Boiling point 72°C
Melting point -141°C
Density 1.559
ND20 1.3909


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR