2,6-Bis(trifluoromethyl)pyridine ≥98% for synthesis

Supplier: Matrix Scientific

007190-1G 007190-5G
101792-152EA 36.92 USD
101792-152 101792-154
2,6-Bis(trifluoromethyl)pyridine ≥98% for synthesis
2,6-Bis(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₇H₃F₆N
Boiling Pt: 82 °C (18mm)
Melting Pt: 55…56 °C
MDL Number: MFCD00236675
CAS Number: 455-00-5

Order Now

Specification Test Results

Boiling point 82°C/18mm
Melting point 55-56°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR