(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
Supplier: Matrix Scientific
Synonyms:
(2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid, (R)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoic acid
007167-250MG
007167-1G
101792-120EA
1600
USD
101792-120
101792-122
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥99%
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid
Hazard code(s): Irritant, Refrigerate, Hygroscopic
Formula:
C₁₀H₉F₃O₃ MW: 234.17 g/mol Boiling Pt: 105…107 °C (1mm) Melting Pt: 43…45 °C |
MDL Number:
MFCD00004184 CAS Number: 20445-31-2 EINECS: 243-829-5 |
Specification Test Results
Boiling point | 105-107°C/1 |
Melting point | 43-45°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...