2-Iodo-3,3,3-trifluoropropene ≥97%

Supplier: Matrix Scientific

007162-1G 007162-5G
101792-108EA 176.92 USD
101792-108 101792-110
2-Iodo-3,3,3-trifluoropropene ≥97%
2-Iodo-3,3,3-trifluoropropene

Hazard code(s): Irritant

Formula: C₃H₂F₃I
Boiling Pt: 66 °C
MDL Number: MFCD02183571
CAS Number: 107070-09-7

Order Now

Specification Test Results

Boiling point 66°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR