2,3,4-Trichlorobenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 2,3,4-Trichlorobenzene-1-sulphonyl chloride

007154-1G 007154-5G
101792-092EA 30.77 USD
101792-092 101792-094
2,3,4-Trichlorobenzenesulfonyl chloride ≥97%
2,3,4-Trichlorobenzenesulphonyl chloride

Hazard code(s): Corrosive, Moisture Sensitive

Formula: C₆H₂Cl₄O₂S
MW: 279.96 g/mol
Melting Pt: 63…65 °C
MDL Number: MFCD00024871
CAS Number: 34732-09-7
EINECS: 252-174-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Melting point 63-65°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR