4-Bromo-2-methylbenzenesulfonyl chloride ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-methylbenzene-1-sulphonyl chloride

007131-1G 007131-5G
101792-076EA 16.92 USD
101792-076 101792-078
4-Bromo-2-methylbenzenesulfonyl chloride ≥97%
4-Bromo-2-methylbenzenesulphonyl chloride

Hazard code(s): Moisture Sensitive, Refrigerate

Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Boiling Pt: 164 °C (15mm)
MDL Number: MFCD00219927
CAS Number: 139937-37-4
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 164°C/15mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR