Nonafluoro-2-(trifluoromethyl)-2-pentene ≥90%

Supplier: Matrix Scientific

Synonyms: Perfluoro(2-methyl-2-pentene), Perfluoro(2-methylpent-2-ene), 1,1,1,3,4,4,5,5,5-Nonafluoro-2-(trifluoromethyl)-2-pentene

007114-25G 007114-100G
101792-048EA 56.92 USD
101792-048 101792-050
Nonafluoro-2-(trifluoromethyl)-2-pentene ≥90%
Nonafluoro-2-(trifluoromethyl)-2-pentene

Contains 5-10% Perfluoro(4-methylpent-2-ene


Hazard code(s): Irritant

Formula: C₆F₁₂
MW: 300.05 g/mol
Boiling Pt: 48…54 °C
Density: 1.62 g/cm³
MDL Number: MFCD00015724
CAS Number: 1584-03-8
EINECS: 216-436-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Boiling point 48-54°C
Density 1.62

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR