2-Chloro-5-nitrocinnamic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-(2-Chloro-5-nitrophenyl)acrylic acid

007070-5G 007070-25G
101791-956EA 27.69 USD
101791-956 101791-958
2-Chloro-5-nitrocinnamic acid ≥98%
2-Chloro-5-nitrocinnamic acid

Hazard code(s): Irritant

Formula: C₉H₆ClNO₄
MW: 227.6 g/mol
Melting Pt: 223 - 234 °C
MDL Number: MFCD00063312
CAS Number: 36015-19-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Melting point 222-223°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR