4-Amino-3-(trifluoromethyl)pyridine ≥99% for synthesis

Supplier: Matrix Scientific

007058-250MG 007058-1G
101791-926EA 107.69 USD
101791-926 101791-928
4-Amino-3-(trifluoromethyl)pyridine ≥99% for synthesis
4-Amino-3-(trifluoromethyl)pyridine

Hazard code(s): Toxic

Formula: C₆H₅F₃N₂
Melting Pt: 101…102 °C
MDL Number: MFCD02183554
CAS Number: 387824-61-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 101-102°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR