2-Amino-4-(trifluoromethyl)pyridine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethyl)pyridin-2-amine, 4-(Trifluoromethyl)-2-pyridylamine

007057-1G 007057-5G
101791-922EA 16.92 USD
101791-922 101791-924
2-Amino-4-(trifluoromethyl)pyridine ≥98% for synthesis
2-Amino-4-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₅F₃N₂
Melting Pt: 72…73 °C
MDL Number: MFCD00077455
CAS Number: 106447-97-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 72-73°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR